missing translation for 'onlineSavingsMsg'
Learn More

Nickel powder, spherical, -400 mesh, 99.5+% (metals basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 4370436

 View more versions of this product

Catalog No. AA4370436


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel powder
500 g
Odorless
(metals basis)
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
58.69
CHEBI:28112
7440-02-0
−400 Mesh
Spherical Powder
Ni
UN3089
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
935
≥99.8%

Chemical Identifiers

7440-02-0
58.69
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137 MFCD06798735
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H228-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Flammable solid.
May cause an allergic skin reaction.

P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c

H228-H317-H350-H372

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

EINECSNumber : 231-111-4

RTECSNumber : QR5950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only