missing translation for 'onlineSavingsMsg'
Learn More

Nickel powder, APS 2.2-3.0 micron, 99.9% (metals basis), C typically <0.25%, Thermo Scientific Chemicals

Catalog No. AA10255A3
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10255A3 2 kg
AA1025536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA10255A3 Supplier Thermo Scientific Chemicals Supplier No. 010255A3

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Density Apparent density 0.5 to 0.65g/cm3
Quantity 2 kg
Odor Odorless
UN Number UN3089
Percent Purity 99.9%
Assay (metals basis), C typically <0.1%
Chemical Name or Material Nickel powder
Hazard Category H228-H317-H350-H372
Hazard Statement GHS H Statement
H372-H351-H228-H317;
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Flammable solid.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 231-111-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only