missing translation for 'onlineSavingsMsg'
Learn More

Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4321414
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4321414 25 g
AA4321422 100 g
AA4321436 500 g
AA43214A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4321414 Supplier Thermo Scientific Chemicals Supplier No. 04321414

Form: Conductive dendritic powder

Conductive surface coatings, inks, adhesives, and sealants

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Silver to Gray
Appearance Silver gray powder
Quantity 25 g
Mesh Size -325 Mesh
Odor Odorless
Form Conductive Dendritic Powder
Assay Percent Range 99.8% (Metals basis)
Chemical Name or Material Nickel
Hazard Category H228-H317-H350-H372
Hazard Statement Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-111-4
RTECSNumber QR5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only