missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) Trifluoromethanesulfonate 96.0+%, TCI America™
$67.11 - $229.29
Chemical Identifiers
| CAS | 60871-84-3 |
|---|---|
| Molecular Formula | C2F6NiO6S2 |
| Molecular Weight (g/mol) | 356.82 |
| MDL Number | MFCD00673740 |
| InChI Key | KVRSDIJOUNNFMZ-UHFFFAOYSA-L |
| Synonym | nickel ii trifluoromethanesulfonate, nickel ii triflate, nickel 2+ ditriflate, trifluoromethanesulfonic acid nickel ii salt, nickel triflate, acmc-209mmw, ni otf 2, nickelous trifluoromethanesulfonate, nickel 2+ ion ditriflate |
| PubChem CID | 9820095 |
| IUPAC Name | nickel(2+) ditrifluoromethanesulfonate |
| SMILES | [Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
Chemical Identifiers
| 60871-84-3 | |
| 356.82 | |
| KVRSDIJOUNNFMZ-UHFFFAOYSA-L | |
| 9820095 | |
| [Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F |
| C2F6NiO6S2 | |
| MFCD00673740 | |
| nickel ii trifluoromethanesulfonate, nickel ii triflate, nickel 2+ ditriflate, trifluoromethanesulfonic acid nickel ii salt, nickel triflate, acmc-209mmw, ni otf 2, nickelous trifluoromethanesulfonate, nickel 2+ ion ditriflate | |
| nickel(2+) ditrifluoromethanesulfonate |
Specifications
| 60871-84-3 | |
| MFCD00673740 | |
| 1759 | |
| KVRSDIJOUNNFMZ-UHFFFAOYSA-L | |
| nickel(2+) ditrifluoromethanesulfonate | |
| 9820095 | |
| ≥96.0% (T) | |
| Nickel(II) Trifluoromethanesulfonate |
| C2F6NiO6S2 | |
| 1 g | |
| nickel ii trifluoromethanesulfonate, nickel ii triflate, nickel 2+ ditriflate, trifluoromethanesulfonic acid nickel ii salt, nickel triflate, acmc-209mmw, ni otf 2, nickelous trifluoromethanesulfonate, nickel 2+ ion ditriflate | |
| [Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F | |
| 356.82 | |
| 356.82 | |
| Crystalline Powder |
Safety and Handling
TSCA : No