accessibility menu, dialog, popup

UserName

CAS RN 60871-84-3

IUPAC Name: nickel(2+) ditrifluoromethanesulfonate
Synonyms: nickel(2+) ditriflate
Molecular Weight (g/mol): 356.82
Molecular Formula: C2F6NiO6S2
InChi Key: KVRSDIJOUNNFMZ-UHFFFAOYSA-L
SMILES: [Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)