missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Trifluoromethanesulfonate 96.0+%, TCI America™

Catalog No. N08611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08611G 1 g
N08615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08611G Supplier TCI America Supplier No. N08611G

Chemical Identifiers

CAS 60871-84-3
Molecular Formula C2F6NiO6S2
Molecular Weight (g/mol) 356.82
MDL Number MFCD00673740
InChI Key KVRSDIJOUNNFMZ-UHFFFAOYSA-L
Synonym nickel ii trifluoromethanesulfonate, nickel ii triflate, nickel 2+ ditriflate, trifluoromethanesulfonic acid nickel ii salt, nickel triflate, acmc-209mmw, ni otf 2, nickelous trifluoromethanesulfonate, nickel 2+ ion ditriflate
PubChem CID 9820095
IUPAC Name nickel(2+) ditrifluoromethanesulfonate
SMILES [Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 1 g
UN Number 1759
Formula Weight 356.82
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nickel(II) Trifluoromethanesulfonate