missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) trifluoromethanesulfonate, 96%, Thermo Scientific Chemicals

Catalog No. AAH6087803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6087803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6087803 Supplier Thermo Scientific Chemicals Supplier No. H6087803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 60871-84-3 | Molecular Formula: C2F6NiO6S2 | Molecular Weight: 356.82 g/mol

It is used as a pharmaceutical intermediate and as OLED chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and as OLED chemicals.

Solubility
Partly soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 60871-84-3
Molecular Formula C2F6NiO6S2
Molecular Weight (g/mol) 356.82
MDL Number MFCD00673740
InChI Key KVRSDIJOUNNFMZ-UHFFFAOYSA-L
Synonym nickel ii trifluoromethanesulfonate, nickel ii triflate, nickel 2+ ditriflate, trifluoromethanesulfonic acid nickel ii salt, nickel triflate, acmc-209mmw, ni otf 2, nickelous trifluoromethanesulfonate, nickel 2+ ion ditriflate
PubChem CID 9820095
IUPAC Name nickel(2+);trifluoromethanesulfonate
SMILES [Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point 100°C to 106°C
Linear Formula Ni(OSO2CF3)2
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Partly soluble in water.
Formula Weight 356.83
Percent Purity 96%
Chemical Name or Material Nickel(II) trifluoromethanesulfonate
Hazard Category H315-H317-H319-H335-H350
Hazard Statement GHS H Statement
H350-H315-H319-H317-H335
May cause cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P272-P280-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only