missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) tetrafluoroborate hexahydrate, 99%

Catalog No. p-43648
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC358730100 10 g Glass bottle
AC358730250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC358730100 Supplier Thermo Scientific Chemicals Supplier No. 358730100

CAS: 15684-36-3 | B2F8H12NiO6 | 340.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 15684-36-3 , 14708-14-6
Molecular Formula B2F8H12NiO6
Molecular Weight (g/mol) 340.39
MDL Number MFCD00150259
InChI Key BFFWGFMAXHCLDV-UHFFFAOYSA-N
Synonym nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate
PubChem CID 177615
SMILES O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F

Specifications

Color Blue-Green
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystals
Quantity 10 g
Assay Percent Range 98.5% min. (Complexometry)
Linear Formula Ni(BF4)2·6H2O
Solubility Information Solubility in water: soluble in water
IUPAC Name nickel(2+) bis(tetrafluoroboranuide) hexahydrate
Formula Weight 340.39
Percent Purity 99%
Packaging Glass bottle
Chemical Name or Material Nickel(II) tetrafluoroborate hexahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
May cause cancer.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 238-753-4
RTECSNumber QR6650000
TSCA TSCA

RUO – Research Use Only