accessibility menu, dialog, popup

UserName

CAS RN 15684-36-3

H2OH2OH2OH2OH2OH2ONiFBFFFFBFFF

CAS RN 15684-36-3

IUPAC Name: nickel(2+) bis(tetrafluoroboranuide) hexahydrate
Synonyms: nickel(2+) hexahydrate ditetrafluoroborate
Molecular Weight (g/mol): 340.39
Molecular Formula: B2F8H12NiO6
InChi Key: BFFWGFMAXHCLDV-UHFFFAOYSA-N
SMILES: O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)