missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, 97%, green, reagent, powder

Catalog No. p-43733
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC415581000 100 g Glass bottle
AC415585000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC415581000 Supplier Thermo Scientific Chemicals Supplier No. 415581000
Only null left

CAS: 1313-99-1 | NiO | 74.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Melting Point 1960°C
Color Green
CAS Min % 96.0
CAS Max % 100.0
Physical Form Powder
Quantity 100 g
Assay Percent Range 96% min. (Complexometry)
Linear Formula NiO
Merck Index 15, 6597
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in caustic solutions,soluble in acids and ammonium hydroxide
Formula Weight 74.7
Percent Purity 97%
Packaging Glass bottle
Specific Gravity 6.67
Density 6.6700g/mL
Chemical Name or Material Nickel(II) oxide, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer by inhalation.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
EINECSNumber 215-215-7
RTECSNumber QR8400000
TSCA TSCA

RUO – Research Use Only