missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxide, Puratronic™, 99.998% (metals basis)

Catalog No. AA1081922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1081922 100 g
AA1081906 5 g
AA1081914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1081922 Supplier Thermo Scientific Chemicals Supplier No. 01081922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1313-99-1 | NiO | 74.69 g/mol

Nickel (II) oxide is used in the preparation of nickel alloys including nickel steel alloys, manufacture of glass and porcelain paints. Further nickel oxide is the main component in nickel-iron battery (Edison battery), nickel-cadmium rechargeable battery, and also used in ceramic industry for making frits and porcelain glazes. It also acts as a hydrogenation catalyst. NiO/CNTs (nickel oxide/carbon nanotubes) could be a potential cathode catalyst for oxygen reduction reaction (ORR) in microbial fuel cells (MFCs).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1313-99-1
Molecular Formula NiO
Molecular Weight (g/mol) 74.69
MDL Number MFCD00011145
InChI Key GNRSAWUEBMWBQH-UHFFFAOYSA-N
Synonym nickel ii oxide, nickel monoxide, bunsenite, nickelous oxide, nickel oxide, mononickel oxide, nickel protoxide, black nickel oxide, nickel 2+ oxide
PubChem CID 14805
IUPAC Name oxonickel
SMILES O=[Ni]

Specifications

Melting Point 1984°C
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99% (metals basis)
Density 6.67
Solubility Information Soluble in ammonium hydroxide. Insoluble in water.
Formula Weight 74.71
Chemical Name or Material Nickel(II) oxide
Hazard Category H317-H350i-H372
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 215-215-7
RTECSNumber QR8400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only