missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) oxalate dihydrate

Catalog No. p-7050785
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3945418 50 g
AA3945430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3945418 Supplier Thermo Scientific Chemicals Supplier No. 03945418

CAS: 6018-94-6 | C2H4NiO6 | 182.741 g/mol

It is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6018-94-6
Molecular Formula C2H4NiO6
Molecular Weight (g/mol) 182.741
MDL Number MFCD00167155
InChI Key VNYOIRCILMCTHO-UHFFFAOYSA-L
Synonym nickel oxalate, nickel ii oxalate dihydrate, nickel oxalate dihydrate, acmc-1b92x, nickel 2+ ; oxalate; dihydrate, nickel oxalate,, nickel 2+ dihydrate oxalate, nickel 2+ ion dihydrate oxalate
PubChem CID 516789
IUPAC Name nickel(2+);oxalate;dihydrate
SMILES C(=O)(C(=O)[O-])[O-].O.O.[Ni+2]

Specifications

Odor Odorless
Quantity 50 g
UN Number UN3077
Merck Index 14,6514
Solubility Information Soluble in acids; Insoluble in water
Formula Weight 182.76 (146.73 Anhydrous)
Physical Form Powder
Chemical Name or Material Nickel(II) oxalate dihydrate
Hazard Category H302+H312-H334-H350-H372
Hazard Statement GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P312-P330-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 208-933-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only