accessibility menu, dialog, popup

UserName

CAS RN 6018-94-6

IUPAC Name: nickel(2+) dihydrate oxalate
Synonyms: nickel(2+) dihydrate oxalate
Molecular Weight (g/mol): 182.74
Molecular Formula: C2H4NiO6
InChi Key: VNYOIRCILMCTHO-UHFFFAOYSA-L
SMILES: O.O.[Ni++].[O-]C(=O)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results