missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) Dibutyldithiocarbamate 97.0+%, TCI America™
$65.24 - $301.98
Chemical Identifiers
| CAS | 13927-77-0 |
|---|---|
| Molecular Formula | C18H36N2NiS4 |
| Molecular Weight (g/mol) | 467.433 |
| MDL Number | MFCD00067273 |
| InChI Key | HPOWMHUJHHIQGP-UHFFFAOYSA-L |
| Synonym | Dibutyldithiocarbamic Acid Nickel(II) Salt, Nickel(II) Bis(dibutyldithiocarbamate) |
| PubChem CID | 6101499 |
| IUPAC Name | N,N-dibutylcarbamodithioate;nickel(2+) |
| SMILES | CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Ni+2] |
Chemical Identifiers
| 13927-77-0 | |
| 467.433 | |
| HPOWMHUJHHIQGP-UHFFFAOYSA-L | |
| 6101499 | |
| CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Ni+2] |
| C18H36N2NiS4 | |
| MFCD00067273 | |
| Dibutyldithiocarbamic Acid Nickel(II) Salt, Nickel(II) Bis(dibutyldithiocarbamate) | |
| N,N-dibutylcarbamodithioate;nickel(2+) |
Specifications
| 13927-77-0 | |
| C18H36N2NiS4 | |
| 25 g | |
| HPOWMHUJHHIQGP-UHFFFAOYSA-L | |
| N,N-dibutylcarbamodithioate;nickel(2+) | |
| 6101499 | |
| ≥97.0% (T) | |
| Nickel(II) Dibutyldithiocarbamate |
| 89°C | |
| MFCD00067273 | |
| Dibutyldithiocarbamic Acid Nickel(II) Salt, Nickel(II) Bis(dibutyldithiocarbamate) | |
| CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Ni+2] | |
| 467.433 | |
| 467.43 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1247
RTECSNumber : QR6140000
TSCA : Yes