missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Dibutyldithiocarbamate 97.0+%, TCI America™

Supplier:  TCI America D1781500G

 View more versions of this product

Catalog No. D1781500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Nickel(II) Dibutyldithiocarbamate
89°C
MFCD00067273
Dibutyldithiocarbamic Acid Nickel(II) Salt, Nickel(II) Bis(dibutyldithiocarbamate)
CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Ni+2]
467.433
467.43
Crystalline Powder
13927-77-0
C18H36N2NiS4
500 g
HPOWMHUJHHIQGP-UHFFFAOYSA-L
N,N-dibutylcarbamodithioate;nickel(2+)
6101499
≥97.0% (T)

Chemical Identifiers

13927-77-0
467.433
HPOWMHUJHHIQGP-UHFFFAOYSA-L
6101499
CCCCN(CCCC)C(=S)[S-].CCCCN(CCCC)C(=S)[S-].[Ni+2]
C18H36N2NiS4
MFCD00067273
Dibutyldithiocarbamic Acid Nickel(II) Salt, Nickel(II) Bis(dibutyldithiocarbamate)
N,N-dibutylcarbamodithioate;nickel(2+)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1247

RTECSNumber : QR6140000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions