missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) citrate hydrate, 98%

Catalog No. AA3963422
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3963422 100 g
AA3963436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3963422 Supplier Thermo Scientific Chemicals Supplier No. 03963422

CAS: 6018-92-4 | C12H10Ni3O14 | 554.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6018-92-4
Molecular Formula C12H10Ni3O14
Molecular Weight (g/mol) 554.28
MDL Number MFCD00054366
InChI Key UPPLJLAHMKABPR-UHFFFAOYSA-H
Synonym nickel ii citrate hydrate
PubChem CID 131881812
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid;nickel
SMILES [Ni++].[Ni++].[Ni++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O

Specifications

Quantity 100 g
UN Number UN3077
Formula Weight 554.33 (Anhydrous)
Percent Purity ≥98%
Physical Form Chunks and/or Powder
Chemical Name or Material Nickel(II) citrate hydrate
Hazard Category H302-H317-H334-H341-H350-H360-H372
Hazard Statement GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 227-873-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only