accessibility menu, dialog, popup

UserName

CAS RN 6018-92-4

IUPAC Name: trinickel(2+) bis(2-hydroxypropane-1,2,3-tricarboxylate)
Synonyms: trinickel(2+) dicitrate
Molecular Weight (g/mol): 554.28
Molecular Formula: C12H10Ni3O14
InChi Key: UPPLJLAHMKABPR-UHFFFAOYSA-H
SMILES: [Ni++].[Ni++].[Ni++].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O.OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results