missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) chloride hexahydrate, 99.999%, (trace metal basis)

CAS: 7791-20-0 | Cl2Ni·6H2O | 237.71 g/mol

Supplier:  Thermo Scientific Chemicals 193570050

 View more versions of this product

Catalog No. AC193570050


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

Nickel(II) chloride hexahydrate
100.0
Solution
99.999%
NiCl2·6H2O
14,351; 15,96; 17,86
Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol
237.71
Trace Metal Basis
7791-20-0,7718-54-9
100.0
5 g
Cl2Ni·6H2O
MFCD00149809
15, 6590
237.71
99.999%
Glass bottle

Chemical Identifiers

7791-20-0
Cl2Ni·6H2O
237.71
MFCD00149809
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing genetic defects.
Very toxic to aquatic life with long lasting effects.<br

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid relea

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only