missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) chloride hexahydrate, 99.999%, (trace metal basis)
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
Specifications
Specifications
| CAS Min % | 100.0 |
| CAS Max % | 100.0 |
| Physical Form | Solution |
| Quantity | 5 g |
| Assay Percent Range | 99.999% |
| Linear Formula | NiCl2·6H2O |
| Fieser | 14,351; 15,96; 17,86 |
| Merck Index | 15, 6590 |
| Solubility Information | Solubility in water: 2540g/L water (20°C). Other solubilities: soluble in ammonium hydroxide, soluble in alcohol |
| Formula Weight | 237.71 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Toxic if swallowed. Toxic if inhaled. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. Very toxic to aquatic life with long lasting effects.<br |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Wear eye protection/face protection. Obtain special instructions before use. IF exposed or concerned: Get medical advice/attention. Avoid relea |
RUO – Research Use Only