missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) 2-Amino-5-methylbenzenesulfonate 95.0+%, TCI America™

Catalog No. a043025g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A043025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A043025G Supplier TCI America Supplier No. A043025G
Only null left

Chemical Identifiers

CAS 375387-13-6
Molecular Formula C14H16N2NiO6S2
Molecular Weight (g/mol) 431.103
MDL Number MFCD00135316
InChI Key JFVHHXOSEDMKHN-UHFFFAOYSA-L
Synonym 4-Aminotoluene-3-sulfonic Acid Nickel(II) Salt, 4-Methylaniline-2-sulfonic Acid Nickel(II) Salt, Nickel(II) 4-Aminotoluene-3-sulfonate, Nickel(II) 4-Methylaniline-2-sulfonate, p-Toluidine-2-sulfonic Acid Nickel(II) Salt, 2-Amino-5-methylbenzenesulfonic Ac
PubChem CID 53384453
IUPAC Name 2-amino-5-methylbenzenesulfonate;nickel(2+)
SMILES CC1=CC(=C(C=C1)N)S(=O)(=O)[O-].CC1=CC(=C(C=C1)N)S(=O)(=O)[O-].[Ni+2]

Specifications

Color Green
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 431.10
Percent Purity ≥95.0% (T)
Chemical Name or Material Nickel(II) 2-Amino-5-methylbenzenesulfonate