missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) 2,4-pentanedionate, 95%

Catalog No. p-7049700
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1881106 5 g
AA1881114 25 g
AA1881122 100 g
AA1881136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1881106 Supplier Thermo Scientific Chemicals Supplier No. 01881106
Only null left

Catalyst and U.V. stabilizer | CAS: 3264-82-2 | C10H14NiO4 | 256.91 g/mol

Catalyst and U.V. stabilizer.Nickel(II) 2,4-pentanedionate is used as a precursor to nickel bis(cyclooctadiene) catalyst. It is also used in the deposition of nickel(II) oxide thin film by sol-gel techniques on conductive glass substrates. Further, it is used in organic synthesis to produce organometals. It is associated with dimethylgold(III) acetylacetonate is used in gold on nickel plating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3264-82-2
Molecular Formula C10H14NiO4
Molecular Weight (g/mol) 256.91
MDL Number MFCD00000024
InChI Key BMGNSKKZFQMGDH-FDGPNNRMSA-L
Synonym nickel ii acetylacetonate, bis 2,4-pentanedionato nickel ii hydrate, acetylacetone nickel ii salt, bis 2,4-pentanedionato nickel ii
PubChem CID 53384569
SMILES [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point ∼230°C (decomposition)
Color Green
Odor Odorless
Quantity 5 g
Beilstein 4157970
Sensitivity Hygroscopic
Merck Index 14,6500
Solubility Information Soluble in water,alcohol,toluene.
IUPAC Name nickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Formula Weight 256.91
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Nickel(II) 2,4-pentanedionate
Show More Show Less
Hazard Category H302-H317-H334-H350i-H500
Hazard Statement GHS H Statement
H350-H302-H317
May cause cancer.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P341-P308+P313-P330-P333+P313-P342+P311-P363-P501c
EINECSNumber 221-875-7
RTECSNumber SA2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only