accessibility menu, dialog, popup

UserName

CAS RN 3264-82-2

IUPAC Name: nickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Synonyms: nickel(2+) bis((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 256.91
Molecular Formula: C10H14NiO4
InChi Key: BMGNSKKZFQMGDH-FDGPNNRMSA-L
SMILES: [Ni++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Percent Purity 
  • (3)
  • (4)
  • (3)
Physical Form 
  • (3)
  • (3)
  • (4)
Quantity 
  • (3)
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (7)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)