missing translation for 'onlineSavingsMsg'
Learn More

Nickel foil, 2.0mm (0.08in) thick, Puratronic™, 99.9945% (metals basis)

Catalog No. p-7048910
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 x 25 mm
50 x 50 mm
Catalog No. Quantity
AA12042FF 25 x 25 mm
AA12042FI 50 x 50 mm
2 options

Catalog No. AA12042FF

Supplier: Thermo Scientific Chemicals 012042FF

Only null left

CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 25 x 25 mm
Odor Odorless
Weight ≈11.1g/25x25mm
Form Foil
Percent Purity 99.9945%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Nickel foil, 2.0mm (0.08 in.) thick
Hazard Category H317-H351-H372
Hazard Statement GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 231-111-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only