missing translation for 'onlineSavingsMsg'
Learn More

Nickel foil, 0.25mm (0.01in) thick, Puratronic™, 99.994% (metals basis)

Catalog No. p-4824830
Change view
Click to view available options
Quantity:
25 x 50 mm
50 x 100 mm
100 x 250 mm
Catalog No. Quantity
AA12045FG 25 x 50 mm
AA12045FY 50 x 100 mm
AA12045HL 100 x 250 mm
3 options

Catalog No. AA12045FG

Supplier: Thermo Scientific Chemicals 012045FG

Only null left

Form: Approx. 2.8g/25 x 50mm | CAS: 7440-02-0 | Ni | 58.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Color Silver to Gray
Appearance Silver gray foil
Quantity 25 x 50 mm
Odor Odorless
Form Foil
Assay Percent Range 99.994% (Metals basis)
Chemical Name or Material Nickel
Hazard Category H317-H351-H372
Precautionary Statement P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 231-111-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only