Learn More
Nickel Chloride Hexahydrate (Crystalline/Certified), Fisher Chemical™
Supplier: Thermo Fisher Scientific N543
Specifications
| Nickel Chloride Hexahydrate | |
| 7791-20-0 | |
| Solid | |
| ≥97 % | |
| MFCD00149809 | |
| Nickel (II) Chloride Hexahydrate | |
| ≥97% | |
| Pass Test | |
| 1mmHg at 615.6°C |
| Green | |
| 4 to 6 | |
| 3 kg | |
| Cl2Ni · 6 H2O | |
| 3288 | |
| 237.69 | |
| Certified | |
| Glass Bottle | |
| >140°C |
Chemical Identifiers
| 7791-20-0 | |
| Cl2Ni · 6 H2O | |
| MFCD00149809 | |
| Nickel (II) Chloride Hexahydrate |
Safety and Handling
WARNING!
Emergency Overview
Cancer hazard. May cause harm to the unborn child. May cause allergic respiratory and skin reaction. Irritating to skin. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Do not get in eyes, on skin, or on clothing. Avoid breathing vapors or mists. Do not ingest. Use only in area provided with appropriate exhaust ventilation. Use only in well-ventilated areas. Use only in well-ventilated areas. Minimize dust generation and accumulation.
NFPA
Health:2
Flammability:0
Instability:0
DOTInformation : DOT Class 6.1, : Poison