missing translation for 'onlineSavingsMsg'
Learn More

Nickel Chloride Hexahydrate (Crystalline/Certified), Fisher Chemical™

Catalog No. p-32671
Change view
Click to view available options
Quantity:
250 g
500 g
3 kg
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
N54250 250 g Glass Bottle
N54500 500 g Glass Bottle
N543 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N54250 Supplier Thermo Fisher Scientific Supplier No. N54250
Only null left

Chemical Identifiers

CAS 7791-20-0
Molecular Formula Cl2Ni · 6 H2O
MDL Number MFCD00149809
Synonym Nickel (II) Chloride Hexahydrate

Specifications

Color Green
pH 4 to 6
Physical Form Solid
Quantity 250 g
Assay Percent Range ≥97 %
UN Number 3288
Formula Weight 237.69
Percent Purity ≥97%
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 1mmHg at 615.6°C
Decomposition Information >140°C
Chemical Name or Material Nickel Chloride Hexahydrate
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Cancer hazard. May cause harm to the unborn child. May cause allergic respiratory and skin reaction. Irritating to skin. Danger of serious damage to health by prolonged exposure. Possible risks of irreversible effects. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Do not get in eyes, on skin, or on clothing. Avoid breathing vapors or mists. Do not ingest. Use only in area provided with appropriate exhaust ventilation. Use only in well-ventilated areas. Use only in well-ventilated areas. Minimize dust generation and accumulation.

NFPA

Health:2
Flammability:0
Instability:0
DOTInformation DOT Class 6.1, : Poison