missing translation for 'onlineSavingsMsg'
Learn More

Nickel Carbonate, Basic, Tetrahydrate, Powder, 45%, Spectrum™ Chemical

$298.00 - $10753.23

Chemical Identifiers

CAS 39430-27-8
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611945
View Documents
Spectrum Chemical Mfg Cor
N104525GM
25 g Poly Bottle
Each for $298.00
Only null left
 
18611946
View Documents
Spectrum Chemical Mfg Cor
N1045125GM
125 g Poly Bottle
Each for $525.17
Only null left
 
18611947
View Documents
Spectrum Chemical Mfg Cor
N1045500GM
500 g Poly Bottle
Each for $1,215.93
Only null left
 
18611948
View Documents
Spectrum Chemical Mfg Cor
N104525KG
2.5 kg Poly Pail
Each for $4,663.93
Only null left
 
18611949
View Documents
Spectrum Chemical Mfg Cor
N104512KG
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

39430-27-8

Specifications

39430-27-8
25 g
Reagent
1
45%
Poly Bottle
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.