missing translation for 'onlineSavingsMsg'
Learn More

Nickel Carbonate, Basic, Tetrahydrate, Powder, 45%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor N104525KG

 View more versions of this product

Catalog No. 18611948


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

39430-27-8
2.5 kg
Reagent
1
45%
Poly Pail

Chemical Identifiers

39430-27-8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.