Learn More
Nickel aluminum, 99.0% min (metals basis)
CAS: 12003-78-0 | AlNi | 85.675 g/mol
$86.49 - $1006.22
Chemical Identifiers
| CAS | 12003-78-0 |
|---|---|
| Molecular Formula | AlNi |
| Molecular Weight (g/mol) | 85.675 |
| MDL Number | MFCD00167392 |
| InChI Key | NPXOKRUENSOPAO-UHFFFAOYSA-N |
| Synonym | aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel |
| PubChem CID | 6336846 |
| IUPAC Name | aluminum;nickel |
| SMILES | [Al].[Ni] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA2289009
|
Thermo Scientific Chemicals
02289009 |
10 g |
Each for $86.49
|
|
|||||
|
AA2289018
|
Thermo Scientific Chemicals
02289018 |
50 g |
Each for $231.26
|
|
|||||
|
AA2289030
|
Thermo Scientific Chemicals
02289030 |
250 g |
Each for $1,006.22
|
|
|||||
Description
Nickel aluminum is used in strengthening constituent in high-temperature nickel-base superalloys, coating blades in gas turbines and jet engines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 12003-78-0 | |
| 85.675 | |
| NPXOKRUENSOPAO-UHFFFAOYSA-N | |
| 6336846 | |
| [Al].[Ni] |
| AlNi | |
| MFCD00167392 | |
| aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel | |
| aluminum;nickel |
Specifications
| Gray | |
| Gray powder, Intermetallic | |
| Odorless | |
| Powder | |
| MFCD00167392 | |
| Insoluble in water. | |
| [Al].[Ni] | |
| 85.675 | |
| Nickel aluminum |
| 12003-78-0 | |
| 10 g | |
| 99.0% min. (metals basis) | |
| AlNi | |
| aluminum-nickel, nickel aluminum, nickel-aluminum alloy, aluminum-nickel alloy, nickel-aluminum, aluminum-nickel catalyst, al.ni, aluminum, compd. with nickel 1:1, aluminum-nickel, aluminum nickel | |
| NPXOKRUENSOPAO-UHFFFAOYSA-N | |
| aluminum;nickel | |
| 6336846 |
Safety and Handling
P201-P202-P223-P231+P232-P281-P308+P313-P335+P334-P370+P378q-P501c
H261-H350
DOTInformation : Hazard Class: 4.3; Packaging Group: II
EINECSNumber : 234-439-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only