accessibility menu, dialog, popup

UserName

CAS RN 12003-78-0

IUPAC Name: aluminium nickel
Synonyms: aluminium nickel
Molecular Weight (g/mol): 85.67
Molecular Formula: AlNi
InChi Key: NPXOKRUENSOPAO-UHFFFAOYSA-N
SMILES: [Al].[Ni]
Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)