missing translation for 'onlineSavingsMsg'
Learn More

Nickel, AAS standard solution, Specpure™ Ni 1000μg/mL

Catalog No. p-7080422
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA88082AE 100 mL
AA88082AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA88082AE Supplier Thermo Scientific Chemicals Supplier No. 088082AE
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

MDL Number MFCD00011137

Specifications

Physical Form Liquid
Concentration Ni in 5% HNO3
UN Number UN3264
Color Colorless
Quantity 100 mL
Grade Specpure
Chemical Name or Material Nickel, AAS standard solution, Ni 1000μg/mL
Hazard Category H290-H314-H335-H350i-H360D
Hazard Statement GHS H Statement
H314-H317-H208
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only