Learn More
Nickel, 99.9%, (trace metal basis), powder, -325 mesh, Thermo Scientific Chemicals
$74.85 - $74.85
Chemical Identifiers
| CAS | 7440-02-0 |
|---|---|
| Molecular Formula | Ni |
| Molecular Weight (g/mol) | 58.69 |
| MDL Number | MFCD00011137 MFCD06798735 |
| InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
| Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
| PubChem CID | 935 |
| ChEBI | CHEBI:28112 |
| IUPAC Name | nickel |
| SMILES | [Ni] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC193610250
|
Thermo Scientific Chemicals
193610250 |
25 g | Plastic bottle |
Each for $74.85
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
| 7440-02-0 | |
| 58.69 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| 935 | |
| nickel |
| Ni | |
| MFCD00011137 MFCD06798735 | |
| raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
| CHEBI:28112 | |
| [Ni] |
Specifications
| Brown-Gray | |
| 7440-02-0 | |
| 99.85 | |
| 8.9 | |
| Plastic bottle | |
| 99.9% | |
| MFCD00011137 MFCD06798735 | |
| 15, 6583 | |
| PXHVJJICTQNCMI-UHFFFAOYSA-N | |
| nickel | |
| 935 | |
| 58.7 | |
| Trace Metal Basis |
| 1455.0°C | |
| 2730.0°C | |
| 100.0 | |
| 25 g | |
| Fine Powder | |
| Ni | |
| 12,335; 13,197; 17,296 | |
| raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
| [Ni] | |
| 58.69 | |
| CHEBI:28112 | |
| 99.9% | |
| Nickel, 99.9%, -325 mesh |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
May cause an allergic skin reaction.
GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 231-111-4
RTECSNumber : QR5950000
TSCA : TSCA
RUO – Research Use Only