Learn More
Innovatek Nichrome Innoculating Loops
Innovatek Nichrome Innoculating Loops glide smoothly across the agar surface for evenly isolated colonies
Supplier: Innovatek 40303413
Description
- A wide range of premium quality, accurately sized twisted Nichrome wire loops
- Nichrome wire offers all the benefits of platinum at much more reasonable cost
- Accurately formed loops glide smoothly across the agar surface for evenly isolated colonies
- Exacting manufacturing process ensures a perfectly symmetrical loop with equal dimensions, rather than an uneven oval
- Calibration gauge checks that inner diameter remains accurate during daily use
- Consistent and reproducible results
- Accurately calibrated with Evans Blue dye test
- Calibration gauge for quality control in the lab
- Rapid cooling
Specifications
| Reusable | |
| 24 G | |
| 12 Loops | |
| No handle |
| Nichrome Wire | |
| No Handle Included | |
| No |