Learn More
Innovatek Nichrome Innoculating Loops
Description
- A wide range of premium quality, accurately sized twisted Nichrome wire loops
- Nichrome wire offers all the benefits of platinum at much more reasonable cost
- Accurately formed loops glide smoothly across the agar surface for evenly isolated colonies
- Exacting manufacturing process ensures a perfectly symmetrical loop with equal dimensions, rather than an uneven oval
- Calibration gauge checks that inner diameter remains accurate during daily use
- Consistent and reproducible results
- Accurately calibrated with Evans Blue dye test
- Calibration gauge for quality control in the lab
- Rapid cooling
Specifications
Specifications
| Disposable | Reusable |
| Material | Nichrome Wire |
| Gauge | 24 G |
| Length (English) Handle | 8 in. |
| Calibrated | Yes |