missing translation for 'onlineSavingsMsg'
Learn More

Nicardipine hydrochloride, 98%

Catalog No. p-7042070
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6126903 1 g
AAJ6126906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6126903 Supplier Thermo Scientific Chemicals Supplier No. J6126903
Only null left

L-type calcium channel blocker | CAS: 54527-84-3 | C26H30ClN3O6 | 515.991 g/mol

L-type calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54527-84-3
Molecular Formula C26H30ClN3O6
Molecular Weight (g/mol) 515.991
MDL Number MFCD00057327
InChI Key AIKVCUNQWYTVTO-UHFFFAOYSA-N
Synonym nicardipine hydrochloride, cardene, nicardipine hcl, perdipine, nicodel, loxen, angioglebil, bionicard, nicardil, perdipina
PubChem CID 41114
IUPAC Name 5-O-[2-[benzyl(methyl)amino]ethyl] 3-O-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate;hydrochloride
SMILES CC1=C(C(C(=C(N1)C)C(=O)OCCN(C)CC2=CC=CC=C2)C3=CC(=CC=C3)[N+](=O)[O-])C(=O)OC.Cl

Specifications

Color Yellow
Quantity 1 g
UN Number UN2811
Merck Index 14,6495
Formula Weight 515.99
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nicardipine hydrochloride
Hazard Category H301+H311-H330
Hazard Statement GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 259-198-4
RTECSNumber US7972100
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only