accessibility menu, dialog, popup

UserName

HClH3COOH3CHNNOOOONCH3CH3

CAS RN 54527-84-3

IUPAC Name: hydrogen 3-{2-[benzyl(methyl)amino]ethyl} 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate chloride
Synonyms: hydrogen nicardipine chloride
Molecular Weight (g/mol): 515.99
Molecular Formula: C26H30ClN3O6
InChi Key: AIKVCUNQWYTVTO-UHFFFAOYNA-N
SMILES: [H+].[Cl-].COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OCCN(C)CC1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)