missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicardipine hydrochloride, 98%
Description
L-type calcium channel blocker
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| UN Number | UN2811 |
| Merck Index | 14,6495 |
| Formula Weight | 515.99 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | Nicardipine hydrochloride |
Safety and Handling
| missing translation for 'healthHazard1' | H301+H311-H330 |
| missing translation for 'healthHazard2' | GHS H Statement H301-H311-H331 Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. |
| missing translation for 'healthHazard3' | P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P330-P361-P363-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 259-198-4 |
| RTECSNumber | US7972100 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only