missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Niacin, Powder, FCC, 99.5-101%, Spectrum™ Chemical
N1013, 59-67-6, C6H5NO2
Supplier: Spectrum Chemical Mfg Cor N10131KG
Description
Spectrum™ Chemical Niacin, Powder, FCC is a vitamin supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 59-67-6 | |
| 0.001 | |
| 234-238°C | |
| C6H5NO2 | |
| 1 kg | |
| OC(=O)C1=CC=CN=C1 | |
| 123.11 | |
| FCC |
| 1 | |
| 0.01 | |
| Poly Pail | |
| MFCD00006391 | |
| PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
| pyridine-3-carboxylic acid | |
| 99.5 to 101% |
Chemical Identifiers
| 59-67-6 | |
| 123.11 | |
| PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
| OC(=O)C1=CC=CN=C1 |
| C6H5NO2 | |
| MFCD00006391 | |
| pyridine-3-carboxylic acid |