missing translation for 'onlineSavingsMsg'
Learn More

Niacin, Powder, FCC, 99.5-101%, Spectrum™ Chemical

Catalog No. p-7157893
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604776 100 g Poly Bottle
18604777 500 g Poly Bottle
18604778 1 kg Poly Pail
18604779 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604776 Supplier Spectrum Chemical Mfg Cor Supplier No. N1013100GM
Only null left

N1013, 59-67-6, C6H5NO2

Spectrum™ Chemical Niacin, Powder, FCC is a vitamin supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59-67-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD00006391
InChI Key PVNIIMVLHYAWGP-UHFFFAOYSA-N
IUPAC Name pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC=CN=C1

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.01
Melting Range 234-238°C
Packaging Poly Bottle
Quantity 100 g
Percent Purity 99.5 to 101%
Grade FCC