missing translation for 'onlineSavingsMsg'
Learn More

Niacin (Nicotinic Acid) Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014807
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014807 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014807 Supplier Merck Emd Millipore Supplier No. PHR12761G
Pharmaceutical (small molecule)

Specifications

CAS 59-67-6
Flash Point 193°C - Closed Cup
Physical Form Neat
Linear Formula C6H5 NO2
MDL Number MFCD00006391
Melting Point 236°C to 239°C
Synonym Nicotinic acid; 3-Picolinic acid; Niacin; Pellagra preventive factor; Pyridine-3-carboxylic acid; Vitamin B3
Type Certified Reference Material
Quantity 1 g
Formula Weight 123.11
Grade Certified Reference Material
Show More Show Less