missing translation for 'onlineSavingsMsg'
Learn More

Cytiva NHS-Activated Sepharose™ 4 Fast Flow Affinity Media

Catalog No. 45002965
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45002965 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45002965 Supplier Cytiva Supplier No. 17090601

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Use for coupling amino-containing smaller proteins and peptides. Cytiva Life Sciences™ NHS-Activated Sepharose™ 4 Fast Flow Affinity Media yields coupled products that are very stable, especially at high pH.

  • Composed of cross-linked 4% agarose beads, NHS activated for coupling amino-containing proteins and peptides
  • NHS (N-hydroxysuccinimide) coupling forms chemically stable amide bond with ligands containing primary amino groups
  • Resin provides spacer arm to coupled ligand
  • Fast Flow matrix gives high productivity
  • Coupled product is very stable, especially at high pH
  • Available in range of different bulk pack sizes and convenient pre-packed formats for easy scale-up and process development

Specifications

Quantity 25 mL
For Use With (Application) For Coupling Amino-Containing Smaller Proteins and Peptides
Stability Stable in commonly used aqueous Buffers provided that the ligand withstand the pH or the chemical environment
Storage Requirements 2°C to 8°C
Product Type NHS-Activated Sepharose 4 Fast Flow Media
Flow Rate 150 to 250 cm/Hoμr
Particle Size 45 to 165 μm
Height (English) Bed 9.84 in.
Height (Metric) Bed 25 cm
Max. Pressure 100 kPa
Matrix 4% Cross-Linked Agarose
pH Range 3 to 13
Storage Buffer 100% Isopropanol
Ligand Type N-hydroxysuccinimide (NHS) Activated
Show More Show Less