missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ NHP XL Cytokine Luminex™ Performance 37-plex Fixed Panel

Catalog No. LKTM021
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LKTM021 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LKTM021 Supplier R&D Systems™ Supplier No. LKTM021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

NHP Fixed XL Cytokine Panel

Specifications

Quantity 1 Kit
Species Primate
Binding Target Multiplex Cytokine Assay Kits
Product Line R&D Systems
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.