missing translation for 'onlineSavingsMsg'
Learn More

NHI-D FAME Mix (Neat), MilliporeSigma™ Supelco™

Catalog No. 111002592
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002592 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002592 Supplier Merck Emd Millipore Supplier No. O83811AMP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Packaging Ampule
CAS 124-10-7
Shipping Condition Dry Ice
Physical Form Solid
Type Analytical Standard
Quantity 100 mg
Grade Analytical Standard