missing translation for 'onlineSavingsMsg'
Learn More

[NH2Me2][(RuCl((S)-segphos(regR)))2(mu-Cl)3], TCI America™

Catalog No. n1022200mg
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1022200MG 200 mg
N10221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1022200MG Supplier TCI America Supplier No. N1022200MG

Chemical Identifiers

CAS 488809-34-3
Molecular Formula C78H67Cl5NO8P4Ru2+
Molecular Weight (g/mol) 1649.678
MDL Number MFCD09753037
InChI Key GTKMQYLNAZIAEF-UHFFFAOYSA-M
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-5,5′C-bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole]diruthenate(II)
PubChem CID 121233601
IUPAC Name [4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;trichloronioruthenium(1-);chloride;hydrochloride
SMILES CNC.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl.[Cl-].[ClH+][Ru-]([ClH+])[ClH+].[Ru]

Specifications

Quantity 200 mg
Formula Weight 1646.65
Physical Form Crystalline Powder
Chemical Name or Material [NH2Me2][(RuCl((S)-segphos(regR)))2(mu-Cl)3]