missing translation for 'onlineSavingsMsg'
Learn More

[NH2Me2][(RuCl((S)-dm-segphos(regR)))2(mu-Cl)3], TCI America™

Catalog No. N1024200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1024200MG 200 mg
N10241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N1024200MG Supplier TCI America Supplier No. N1024200MG

Chemical Identifiers

CAS 944451-14-3
Molecular Formula C94H99Cl5NO8P4Ru2+
Molecular Weight (g/mol) 1874.11
MDL Number MFCD09753038
InChI Key CABGTUSYWYURQC-UHFFFAOYSA-M
Synonym Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-5,5′C-bis[di(3,5-xylyl)phosphino]-4,4′C-bi-1,3-benzodioxole]diruthenate(II)
PubChem CID 121233602
IUPAC Name [4-[5-bis(3,5-dimethylphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-dimethylphenyl)phosphane;N-methylmethanamine;ruthenium;trichloronioruthenium(1-);chloride;hydrochloride
SMILES CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CC1=CC(=CC(=C1)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.CNC.Cl.[Cl-].[ClH+][Ru-]([ClH

Specifications

Quantity 200 mg
Formula Weight 1871.09
Physical Form Crystalline Powder
Chemical Name or Material [NH2Me2][(RuCl((S)-dm-segphos(regR)))2(mu-Cl)3]