missing translation for 'onlineSavingsMsg'
Learn More

[NH2Me2][(RuCl((S)-binap))2(mu-Cl)3], TCI America™

Supplier:  TCI America N10161G

 View more versions of this product

Catalog No. N10161G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

[NH2Me2][(RuCl((S)-binap))2(mu-Cl)3]
C90H78Cl5NP4Ru2+2
1 g
IDNUVQHPQCZKMV-UHFFFAOYSA-M
trichloranium;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride
132472188
Crystalline Powder
199541-17-8
MFCD09753033
Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]diruthenate(II)
CNC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru]
1676.906
1670.86

Chemical Identifiers

199541-17-8
1676.906
IDNUVQHPQCZKMV-UHFFFAOYSA-M
132472188
CNC.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=CC=C7)C8=CC=CC=C8.[ClH2+].[ClH2+].[ClH2+].Cl.[Cl-].[Ru].[Ru]
C90H78Cl5NP4Ru2+2
MFCD09753033
Dimethylammonium Dichlorotri(mu-chloro)bis[(S)-(-)-2,2′C-bis(diphenylphosphino)-1,1′C-binaphthyl]diruthenate(II)
trichloranium;[1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane;N-methylmethanamine;ruthenium;chloride;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions