missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[NH2Me2][(RuCl((S)-binap))2(mu-Cl)3], TCI America™
Specifications
Specifications
| Quantity | 200 mg |
| Formula Weight | 1670.86 |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | [NH2Me2][(RuCl((S)-binap))2(mu-Cl)3] |
Safety and Handling
| TSCA | No |