missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse NGF Recombinant Protein

Catalog No. A42626
Change view
Click to view available options
Quantity:
5 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42626 5 μg
A42627 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A42626 Supplier Invitrogen™ Supplier No. A42626
Only null left

Recombinant Protein

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/mL or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Nerve growth factor (NGF) influences the survival and differentiation of a variety of neuronal and nonneuronal cells. The actions of this polypeptide result from binding to specific cell surface receptors which are present as both high and low affinity sites. Although both types of receptor bind NGF only the high affinity form is capable of signal transduction. Cross-linking studies with 125I-NGF have shown that the high affinity receptor of rat PC12 cells forms an Mr 158,000 complex with NGF while the low affinity receptor forms an Mr 100,000 complex. Full length cDNA clones for both the human and rat NGF receptor genes have been produced and sequenced, revealing greater than 90% homology between the two species. NGF exhibits nerve growth stimulating activity and is involved in the regulation of growth and the differentiation of sympathetic and certain sensory neurons. Further, NGF stimulates division and differentiation of sympathetic and embryonic sensory neurons. Mutations in the NGF gene have been associated with hereditary sensory and autonomic neuropathy, type 5 (HSAN5), and dysregulation of NGF gene expression is associated with allergic rhinitis.

Specifications

Accession Number P01139
For Use With (Application) Control, Bioactivity
Formulation 50mM sodium acetate with 5% mannitol, 150mM NaCl, 5% trehalose and no preservative; pH 5.5
Gene ID (Entrez) 18049
Molecular Weight (g/mol) 13.5 kDa
Name Mouse NGF
pH Range 5.5
Purification Method Purified
Quantity 5 μg
Storage Requirements -20°C or -80°C if preferred
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias B nerve growth factor; beta ngf; beta-nerve growth factor; BetaNGF; beta-NGF; bNGF; b-NGF; HSAN5; MGC161426; MGC161428; Nerve growth factor; nerve growth factor (beta polypeptide); nerve growth factor, beta; nerve growth factor, beta polypeptide; nerve growth factor, beta subunit; NGF; NGF b; NGF β; Ngfb; NGFbeta; NGFβ; β nerve growth factor; β NGF; βNGF
Common Name NGF
Gene Symbol NGF
Product Type Protein
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Protein Tag Untagged
Sequence Mouse NGF, amino acids Ser122-Gly241 (Accession # NP_001106168.1) with an initial C-terminal Met
Content And Storage Store at -20 to -70 °C
Activity Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. The ED50 for this effect is 1-6 ng/mL.
Endotoxin Level < 1 EU/μg
Shipping Condition Room Temperature
Expression System CHO cells
Protein Family Growth Factors & Receptors
Protein Form Native
Form Lyophilized
Protein Subtype Neurotrophic Factors
Research Category Development, Immunology, Inflammation, Neurobiology
Purity or Quality Grade >96% by SDS-PAGE
Protein nerve growth factor
Show More Show Less