missing translation for 'onlineSavingsMsg'
Learn More

Ngamma-Trityl-L-asparagine hydrate, 98%

CAS: 132388-58-0 | C23H22N2O3 | 374.44 g/mol

$70.62 - $234.78

Chemical Identifiers

CAS 132388-58-0
Molecular Formula C23H22N2O3
Molecular Weight (g/mol) 374.44
MDL Number MFCD00153422
InChI Key BRRPJQYCERAMFI-GNLPSFAGNA-N
Synonym h-asn trt-oh, n'-trityl-l-asparagine, n-beta-trityl-l-asparagine, n-triphenylmethyl-l-asparagine, 2s-2-amino-4-oxo-4-tritylamino butanoic acid, 2s-2-amino-3-triphenylmethylcarbamoyl propanoic acid, 2s-2-amino-3-triphenylmethyl carbamoyl propanoic acid, h-asn trt-2-chlorotrityl resin, asparagine trt, h-asn trt oh
PubChem CID 7408309
IUPAC Name (2S)-2-amino-4-oxo-4-(tritylamino)butanoic acid
SMILES N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6328503
View Documents
Thermo Scientific Chemicals
H6328503
1 g
Each for $70.62
Only null left
 
AAH6328506
View Documents
Thermo Scientific Chemicals
H6328506
5 g
Each for $234.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

132388-58-0
374.44
BRRPJQYCERAMFI-GNLPSFAGNA-N
7408309
N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
C23H22N2O3
MFCD00153422
h-asn trt-oh, n'-trityl-l-asparagine, n-beta-trityl-l-asparagine, n-triphenylmethyl-l-asparagine, 2s-2-amino-4-oxo-4-tritylamino butanoic acid, 2s-2-amino-3-triphenylmethylcarbamoyl propanoic acid, 2s-2-amino-3-triphenylmethyl carbamoyl propanoic acid, h-asn trt-2-chlorotrityl resin, asparagine trt, h-asn trt oh
(2S)-2-amino-4-oxo-4-(tritylamino)butanoic acid

Specifications

132388-58-0
C23H22N2O3
h-asn trt-oh, n'-trityl-l-asparagine, n-beta-trityl-l-asparagine, n-triphenylmethyl-l-asparagine, 2s-2-amino-4-oxo-4-tritylamino butanoic acid, 2s-2-amino-3-triphenylmethylcarbamoyl propanoic acid, 2s-2-amino-3-triphenylmethyl carbamoyl propanoic acid, h-asn trt-2-chlorotrityl resin, asparagine trt, h-asn trt oh
N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O
1 g
7408309
98%
Ngamma-Trityl-L-asparagine hydrate
White
MFCD00153422
BRRPJQYCERAMFI-GNLPSFAGNA-N
(2S)-2-amino-4-oxo-4-(tritylamino)butanoic acid
374.44
392.45 (Anhydrous)
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only