missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ngamma-Trityl-L-asparagine hydrate, 98%
CAS: 132388-58-0 | C23H22N2O3 | 374.44 g/mol
Supplier: Thermo Scientific Chemicals H6328506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ngamma-Trityl-L-asparagine hydrate | |
| White | |
| MFCD00153422 | |
| BRRPJQYCERAMFI-GNLPSFAGNA-N | |
| (2S)-2-amino-4-oxo-4-(tritylamino)butanoic acid | |
| 374.44 | |
| 392.45 (Anhydrous) | |
| Powder |
| 132388-58-0 | |
| C23H22N2O3 | |
| h-asn trt-oh, n'-trityl-l-asparagine, n-beta-trityl-l-asparagine, n-triphenylmethyl-l-asparagine, 2s-2-amino-4-oxo-4-tritylamino butanoic acid, 2s-2-amino-3-triphenylmethylcarbamoyl propanoic acid, 2s-2-amino-3-triphenylmethyl carbamoyl propanoic acid, h-asn trt-2-chlorotrityl resin, asparagine trt, h-asn trt oh | |
| N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O | |
| 5 g | |
| 7408309 | |
| 98% |
Chemical Identifiers
| 132388-58-0 | |
| 374.44 | |
| BRRPJQYCERAMFI-GNLPSFAGNA-N | |
| 7408309 | |
| N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O |
| C23H22N2O3 | |
| MFCD00153422 | |
| h-asn trt-oh, n'-trityl-l-asparagine, n-beta-trityl-l-asparagine, n-triphenylmethyl-l-asparagine, 2s-2-amino-4-oxo-4-tritylamino butanoic acid, 2s-2-amino-3-triphenylmethylcarbamoyl propanoic acid, 2s-2-amino-3-triphenylmethyl carbamoyl propanoic acid, h-asn trt-2-chlorotrityl resin, asparagine trt, h-asn trt oh | |
| (2S)-2-amino-4-oxo-4-(tritylamino)butanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only